Learn More
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco™ Injectors
Catalog No. 365GL251
Change view
Click to view available options
Volume (Metric):
50 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
365GL251 50 μL
365JL281 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 365GL251 Supplier Thermo Scientific™ Supplier No. 365GL251
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
Thermo Scientific™ Standard Fixed Needle Syringes for Rheodyne/Valco Injectors offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Specifications
Description 50uL
Needle Gauge 22 G
Length (Metric) 50 mm
Product Type HPLC Autosampler Syringe
Disposable Reusable
Volume (Metric) 50 μL
Needle Point Style 90° Blunt
For Use With (Equipment) Injector
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.