Learn More
Restek™ Stainless-Steel HPLC Column Tubing
Can be used immediately without any additional treatment
Supplier:  Restek™ RK 25117
Encompass
 View more versions of this product
Catalog No. 06712684
Only null left
Description
Description
  • Manufactured from fine, chromatographic-grade Type 316 stainless steel
  • Corrosion-resistant, ultrasonically cleaned and passivated
  • Tubing is polished to ensure a burr-free cut
Specifications
Chemical Identifiers
HPLC Column Tubing
0.25 in.
0.635 cm
Stainless Steel (316)
Specifications
HPLC Column Tubing
0.25 in.
10 in.
Stainless Steel (316)
0.181 in.
0.635 cm
25 cm
Product Suggestions
Product Suggestions