Learn More
OHAUS Stainless Steel Defender 5000 Series Floor Scale
Defender 5000 Series Floor Scales meet advanced weighing needs for industrial and commercial use. Their high-quality construction is designed to minimize flexing under heavy loads for years of dependable performance. The indicator features software for counting, checkweighing, accumulation, display hold and other modes.
Supplier: OHAUS 30837700
Description
- 304 stainless steel, flat surface decks with 4 IP68 load cells, fixed top plate (5 mm/0.196 in. thick) with structural bracing for stiff construction.
- Platforms available in sizes of 3 x 3 ft. and 4 x 4 ft. and capacities from 2,500 lb. to 5,000 lb. with NTEP certified 5,000e performance, allowing greater flexibility for your weighing needs.
- Equipped with a new TD52 series indicator featuring an easy-to-use keypad, large backlit display, and plain text on-screen prompts to guide you through operation.
Specifications
| 1250 kg | |
| LCD | |
| Class III |
| Scale | |
| 0.2 kg | |
| i-DF52XW2500C1L |