Learn More
MilliporeSigma™ Supelco™ Stainless Steel 1/16 in. Capillary Tubing, Length: 5 cm
Use to connect the column to the detector compartment in a capillary gas chromatographic setup
Supplier: MilliporeSigma™ Supelco™ 56719
Chemical Identifiers
| Capillary Tubing | |
| 1/16 in. | |
| Stainless Steel |
| Gas Chromatography (GC) | |
| 5 cm |
Specifications
| Capillary Tubing | |
| 0.010 in. | |
| 5 cm |
| Gas Chromatography (GC) | |
| 1/16 in. | |
| Stainless Steel |