missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stachyose hydrate, technical, Thermo Scientific™
Chemical Identifiers
| CAS | 54261-98-2 |
|---|---|
| Molecular Formula | C24H42O21·xH2O |
| Molecular Weight (g/mol) | 666.58 |
| InChI Key | XQDOUZBIGFWLCL-UHFFFAOYSA-N |
| PubChem CID | 131801001 |
| IUPAC Name | 2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate |
| SMILES | C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)OC4(C(C(C(O4)CO)O)O)CO)O)O)O)O)O)O)O)O)O)O.O.O.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC453142500
|
Thermo Scientific Chemicals
453142500 |
250mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC453140010
|
Thermo Scientific Chemicals
453140010 |
1g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC453140050
|
Thermo Scientific Chemicals
453140050 |
5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 54261-98-2 | |
| 666.58 | |
| 131801001 | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)OC4(C(C(C(O4)CO)O)O)CO)O)O)O)O)O)O)O)O)O)O.O.O.O |
| C24H42O21·xH2O | |
| XQDOUZBIGFWLCL-UHFFFAOYSA-N | |
| 2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate |
Specifications
| 54261-98-2 | |
| C24H42O21·xH2O | |
| + 129.00 (20.00°C c=4, H2O) | |
| Solubility in water: soluble. | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)OC4(C(C(C(O4)CO)O)O)CO)O)O)O)O)O)O)O)O)O)O.O.O.O | |
| 666.58 | |
| 666.58 | |
| Stachyose hydrate |
| Authentic | |
| 250mg | |
| + 129.00 | |
| XQDOUZBIGFWLCL-UHFFFAOYSA-N | |
| 2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate | |
| 131801001 | |
| Technical |
RUO â Research Use Only