missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stachyose hydrate, technical, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 453140010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| Stachyose hydrate | |
| Authentic | |
| 1g | |
| + 129.00 | |
| XQDOUZBIGFWLCL-UHFFFAOYSA-N | |
| 2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate | |
| 131801001 | |
| Technical |
| 54261-98-2 | |
| C24H42O21·xH2O | |
| + 129.00 (20.00°C c=4, H2O) | |
| Solubility in water: soluble. | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)OC4(C(C(C(O4)CO)O)O)CO)O)O)O)O)O)O)O)O)O)O.O.O.O | |
| 666.58 | |
| 666.58 |
Chemical Identifiers
| 54261-98-2 | |
| 666.58 | |
| 131801001 | |
| C(C1C(C(C(C(O1)OCC2C(C(C(C(O2)OCC3C(C(C(C(O3)OC4(C(C(C(O4)CO)O)O)CO)O)O)O)O)O)O)O)O)O)O.O.O.O |
| C24H42O21·xH2O | |
| XQDOUZBIGFWLCL-UHFFFAOYSA-N | |
| 2-[[6-[[6-[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-3,4,5-trihydroxyoxan-2-yl]methoxy]-3,4,5-trihydroxyoxan-2-yl]methoxy]-6-(hydroxymethyl)oxane-3,4,5-triol;trihydrate |
RUO â Research Use Only