missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SRT™ SEC-500 Gel Filtration Guard Column, 5 μm Particle Size, PEEK

Catalog No. 111008008
Change view
Click to view available options
Diameter: 4.6 mm; Length: 5 cm, PEEK
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111008008 Diameter: 4.6 mm; Length: 5 cm, PEEK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111008008 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777058
Only null left

Sepax SRT™ SEC columns feature a hydrophilic surface layer that effectively shields the highly porous, five micron silica particles from interacting with your sample.

SRT™ columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.

Specifications

pH 2.0 to 8.5
Particle Size 5 μm
Length (Metric) 5 cm
Pore Size 500 Å
Type SEC HPLC Guard Column
Diameter (Metric) Inner 4.6 mm
Material PEEK
Phase Size Exclusion
For Use With (Application) HPLC
Product Line SRT

SRT is a registered trademark of Sepax Technologies