missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SRT™ SEC-500 Gel Filtration Column, 5 μm Particle Size, PEEK

Catalog No. 111006240
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111006240 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111006240 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777059

Sepax SRT™ SEC columns feature a hydrophilic surface layer that effectively shields the highly porous, five micron silica particles from interacting with your sample.

SRT™ columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 30 cm
Particle Size 5 μm
pH 2.0 to 8.5
Pore Size 500 Å
Stationary Phase Hydrophilic Polymer
Product Line SRT
Matrix Hydrophilic Polymer Phase/Spherical Silica
Phase Size Exclusion
Material PEEK
Technique SEC HPLC
Show More Show Less

SRT is a registered trademark of Sepax Technologies