missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SRT™ SEC-300 Gel Filtration Guard Column, 5 μm Particle Size, Stainless Steel

Catalog No. 111008510
Change view
Click to view available options
Diameter (Metric) Inner:
4.6 mm
7.8 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner
111008510 4.6 mm
111008511 7.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111008510 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777048

Sepax SRT™ SEC columns feature a hydrophilic surface layer that effectively shields the highly porous, five micron silica particles from interacting with your sample.

SRT™ columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.

Specifications

pH 2.0 to 8.5
Particle Size 5 μm
Length (Metric) 5 cm
Pore Size 300 Å
Type SEC HPLC Guard Column
Diameter (Metric) Inner 4.6 mm
Material Stainless Steel
Phase Size Exclusion
For Use With (Application) HPLC
Product Line SRT

SRT is a registered trademark of Sepax Technologies