missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SRT™ SEC-100 Gel Filtration Guard Column, 5 μm Particle Size, Stainless Steel
Description
SRT™ columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.
Specifications
Specifications
| pH | 2.0 to 8.5 |
| Particle Size | 5 μm |
| Length (Metric) | 5 cm |
| Pore Size | 100 Å |
| Type | SEC HPLC Guard Column |
| Diameter (Metric) Inner | 4.6 mm |
| Material | Stainless Steel |
| Phase | Size Exclusion |
| For Use With (Application) | HPLC |
| Product Line | SRT |
SRT is a registered trademark of Sepax Technologies