missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SRT™ SEC-100 Gel Filtration Column, 5 μm Particle Size, Stainless Steel
Sepax SRT™ SEC columns feature a hydrophilic surface layer that effectively shields the highly porous, five micron silica particles from interacting with your sample.
Supplier: MilliporeSigma™ Supelco™ Z777039
Description
SRT™ columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.Specifications
| 7.8 mm | |
| 5 μm | |
| Hydrophilic Polymer | |
| Hydrophilic Polymer Phase/Spherical Silica | |
| Stainless Steel |
| 30 cm | |
| 100 Å | |
| SRT | |
| Size Exclusion | |
| SEC HPLC |
SRT is a registered trademark of Sepax Technologies