missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SRT™ SEC-100 Gel Filtration Column, 5 μm Particle Size, PEEK
Description
SRT™ columns are employed in the quality control of biotherapeutics by gel filtration chromatography. Select from a wide variety of available pore sizes to analyze peptides, proteins, glycoproteins, vaccines and water soluble polymers.
Specifications
Specifications
| Diameter (Metric) | 4.6 mm |
| Length (Metric) | 30 cm |
| Particle Size | 5 μm |
| Pore Size | 100 Å |
| Stationary Phase | Hydrophilic Polymer |
| Product Line | SRT |
| Matrix | Hydrophilic Polymer Phase/Spherical Silica |
| Phase | Size Exclusion |
| Material | PEEK |
| Technique | SEC HPLC |
SRT is a registered trademark of Sepax Technologies