missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SRT™-C SEC-300 Gel Filtration Guard Column, 5 μm Particle Size, PEEK

Catalog No. 111007725
Change view
Click to view available options
Diameter: 4.6 mm; Length: 5 cm, PEEK
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111007725 Diameter: 4.6 mm; Length: 5 cm, PEEK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111007725 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777111

The bonded phase in SRT™-C SEC columns is more polar than the hydrophilic phase used in SRT™ columns, making SRT™-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies.

SRT™-C columns share with SRT™ columns the same 5 micron silica particle and the same wide range of pore sizes so you can select the pore size that will allow full access for your compound of interest.

Specifications

pH 2.0 to 8.5
Particle Size 5 μm
Length (Metric) 5 cm
Pore Size 300 Å
Type SEC HPLC Guard Column
Diameter (Metric) Inner 4.6 mm
Material PEEK
Phase Size Exclusion
For Use With (Application) HPLC
Product Line SRT

SRT is a registered trademark of Sepax Technologies