missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SRT™-C SEC-2000 Gel Filtration Column, 5 μm Particle Size, Stainless Steel
The bonded phase in SRT™-C SEC columns is more polar than the hydrophilic phase used in SRT™ columns, making SRT™-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies.
$2589.78 - $3137.08
Specifications
| Length (Metric) | 30 cm |
|---|---|
| Particle Size | 5 μm |
| pH | 2.0 to 8.5 |
| Pore Size | 2000 Å |
| Stationary Phase | Hydrophilic Polymer |
Description
SRT™-C columns share with SRT™ columns the same 5 micron silica particle and the same wide range of pore sizes so you can select the pore size that will allow full access for your compound of interest.Specifications
| 30 cm | |
| 2.0 to 8.5 | |
| Hydrophilic Polymer | |
| Hydrophilic Polymer Phase/Spherical Silica | |
| Stainless Steel |
| 5 μm | |
| 2000 Å | |
| SRT | |
| Size Exclusion | |
| SEC HPLC |
SRT is a registered trademark of Sepax Technologies