missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SRT™-C SEC-1000 Gel Filtration Column, 5 μm Particle Size, PEEK

Catalog No. 111006204
Change view
Click to view available options
Length (Metric):
30 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
111006204 30 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111006204 Supplier MilliporeSigma™ Supelco™ Supplier No. Z777124

The bonded phase in SRT™-C SEC columns is more polar than the hydrophilic phase used in SRT™ columns, making SRT™-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies.

SRT™-C columns share with SRT™ columns the same 5 micron silica particle and the same wide range of pore sizes so you can select the pore size that will allow full access for your compound of interest.

Specifications

Diameter (Metric) 4.6 mm
Length (Metric) 30 cm
Particle Size 5 μm
pH 2.0 to 8.5
Pore Size 1000 Å
Stationary Phase Hydrophilic Polymer
Product Line SRT
Matrix Hydrophilic Polymer Phase/Spherical Silica
Phase Size Exclusion
Material PEEK
Technique SEC HPLC
Show More Show Less

SRT is a registered trademark of Sepax Technologies