missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ SRT™-C SEC-100 Gel Filtration Guard Column, 5 μm Particle Size, Stainless Steel
The bonded phase in SRT™-C SEC columns is more polar than the hydrophilic phase used in SRT™ columns, making SRT™-C columns better suited for the gel filtration analysis of more hydrophobic bioopolymers such as membrane proteins and pegylated antibodies.
$950.17 - $950.17
Specifications
| pH | 2.0 to 8.5 |
|---|---|
| Particle Size | 5 μm |
| Length (Metric) | 5 cm |
| Pore Size | 100 Å |
| Type | SEC HPLC Guard Column |
Description
SRT™-C columns share with SRT™ columns the same 5 micron silica particle and the same wide range of pore sizes so you can select the pore size that will allow full access for your compound of interest.Specifications
| 2.0 to 8.5 | |
| 5 cm | |
| SEC HPLC Guard Column | |
| Size Exclusion | |
| SRT |
| 5 μm | |
| 100 Å | |
| Stainless Steel | |
| HPLC |
SRT is a registered trademark of Sepax Technologies