missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SR 11237

Catalog No. 341110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
341110 10 mg
341150 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 341110 Supplier Tocris Bioscience™ Supplier No. 3411/10
Only null left

Pan RXR agonist

Pan retinoid X receptor (RXR) agonist that is devoid of any RAR activity.

Chemical Identifiers

CAS 146670-40-8
Molecular Formula C24H28O4

Specifications

Quantity 10 mg
Formula Weight 380.48
Purity >98%
Molecular Weight (g/mol) 380.484
Chemical Name or Material SR 11237
Recommended Storage Store at -20°C