missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SPOP-i-6lc
Description
SPOP-i-6lc is a selective SPOP E3 ubiquitin ligase inhibitor (IC50 = 2.1 μM and 3.5 μM, in A498 and OS-RC-2 cell lines, respectively). In vitro, SPOP-i-6lc suppresses viability and proliferation of A498 and OS-RC-2 kidney cancer cell lines. SPOP-i-6lc leads to the accumulation of tumor suppressors PTEN and DUSP7 and decreased levels of phosphorylated AKT and ERK in clear-cell renal cell carcinoma (ccRCC) cell lines.
Specifications
Specifications
| Chemical Name or Material | 1,5-Dihydro-2-imino-10-methyl-N-[2-(4-methyl-1-piperazinyl)ethyl]-5-oxo-1-[2-(2-thienyl)ethyl]-2 H-dipyrido[1,2-a:2 ',3 '-d]pyrimidine-3-carboxamide |
| CAS | 2136270-56-7 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Ubiquitin Ligase (E3) Inhibitors |
| Molecular Formula | C26H31N7O2S |
| Purity | 98% |