missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spiro[fluorene-9,9'-xanthene]-3',6'-diol 98.0+%, TCI America™
Supplier: TCI America S09535G
Specifications
| Spiro[fluorene-9,9′-xanthene]-3′,6′-diol | |
| 269°C | |
| MFCD01077103 | |
| ZTQUBBFVYUHQRC-UHFFFAOYSA-N | |
| spiro[fluorene-9,9'-xanthene]-3',6'-diol | |
| 3106818 | |
| ≥98.0% (GC) |
| 4081-00-9 | |
| C25H16O3 | |
| 5 g | |
| OC1=CC2=C(C=C1)C1(C3=CC=CC=C3C3=CC=CC=C13)C1=C(O2)C=C(O)C=C1 | |
| 364.40 | |
| 364.40 | |
| Crystalline Powder |
Chemical Identifiers
| 4081-00-9 | |
| 364.40 | |
| ZTQUBBFVYUHQRC-UHFFFAOYSA-N | |
| spiro[fluorene-9,9'-xanthene]-3',6'-diol |
| C25H16O3 | |
| MFCD01077103 | |
| 3106818 | |
| OC1=CC2=C(C=C1)C1(C3=CC=CC=C3C3=CC=CC=C13)C1=C(O2)C=C(O)C=C1 |
Safety and Handling
TSCA : No