accessibility menu, dialog, popup

UserName

HOOOH

CAS RN 4081-00-9

IUPAC Name: spiro[fluorene-9,9'-xanthene]-3',6'-diol
Synonyms: spiro[fluorene-9,9'-xanthene]-3',6'-diol
Molecular Weight (g/mol): 364.4
Molecular Formula: C25H16O3
InChi Key: ZTQUBBFVYUHQRC-UHFFFAOYSA-N
SMILES: OC1=CC2=C(C=C1)C1(C3=CC=CC=C3C3=CC=CC=C13)C1=C(O2)C=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results