missing translation for 'onlineSavingsMsg'
Learn More

Spiro[fluorene-9,9'-xanthene]-3',6'-diol 98.0+%, TCI America™

Catalog No. S09531G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09531G 1 g
S09535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09531G Supplier TCI America Supplier No. S09531G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4081-00-9
Molecular Formula C25H16O3
Molecular Weight (g/mol) 364.40
MDL Number MFCD01077103
InChI Key ZTQUBBFVYUHQRC-UHFFFAOYSA-N
PubChem CID 3106818
IUPAC Name spiro[fluorene-9,9'-xanthene]-3',6'-diol
SMILES OC1=CC2=C(C=C1)C1(C3=CC=CC=C3C3=CC=CC=C13)C1=C(O2)C=C(O)C=C1

Specifications

Melting Point 269°C
Quantity 1 g
Formula Weight 364.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Spiro[fluorene-9,9′-xanthene]-3′,6′-diol