missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ SPINeasy™ DNA Pro Kit for Soil

Catalog No. MP116546050
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP116546050 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP116546050 Supplier MP Biomedicals™ Supplier No. 116546050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Isolation of microbial DNA from high and low biomass soil samples. Unbiased lysis of microbial populations for more accurate microbiome studies. Inhibitor removal technology extracts nucleic acids free of most notorious PCR inhibitors.

  • The SPINeasy DNA Pro Kit for Soil has been carefully designed for the isolation of pure microbiome genomic DNA from challenging soil types, including those with low biomass or that are highly contaminated. Soil samples are complex environments characterized by the presence of inhibitory compounds, such as humic acid, heavy metals, and other aromatic components, which often prove to be challenging for downstream molecular analysis.
  • The SPINeasy DNA Pro Kit for Soil effectively lyses microbial populations, including bacteria, fungi, viruses, and protists. The kit provides similar yields to that of our highly cited FastDNA™ SPIN Kit, but with improved purity and reduced processing time. Isolated DNA products showed no inhibition in PCR and were immediately ready to be used for downstream applications, including long fragment PCR, qPCR, and next-generation sequencing (16S and whole genome), without the need of further inhibitor removal steps.

Specifications

Content And Storage Store at Room Temperature, Dry Environment
Format Kit
Isolation Technology Spin Column
Sample Type Up to 500 mg of Soil Samples
Includes 50x Lysing Matrix YB, 2 mL RNase A Solution, 55 mL Buffer S1, 22 mL Buffer S2, 50 mL Buffer S3, 28 mL Buffer S4, 40 mL Buffer S5, 6 mL Buffer S6, 50x Column S, 50x 2 mL collection tubes, 50x 1.5 mL collection tubes
Product Type DNA Isolation Kit
Quantity 50 Preps

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.