missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Spin-X™ UF Concentrators

Catalog No. 07201354
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
30 kDa
5 kDa
50 kDa
Quantity:
12/Pk.
25/Pk.
Volume (Metric):
500 μL
6 mL
20 mL
Filtration Area:
0.5 cm2
2.5 cm2
6 cm2
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Quantity Volume (Metric) Filtration Area
07201354 100 kDa 12/Pk. 20 mL 6 cm2
07201340 5 kDa 25/Pk. 500 μL 0.5 cm2
07201342 30 kDa 25/Pk. 500 μL 0.5 cm2
07201341 10 kDa 25/Pk. 500 μL 0.5 cm2
07201343 50 kDa 25/Pk. 500 μL 0.5 cm2
07201349 100 kDa 25/Pk. 6 mL 2.5 cm2
07201345 5 kDa 25/Pk. 6 mL 2.5 cm2
07201346 10 kDa 25/Pk. 6 mL 2.5 cm2
07201344 100 kDa 25/Pk. 500 μL 0.5 cm2
07201348 50 kDa 25/Pk. 6 mL 2.5 cm2
07201347 30 kDa 25/Pk. 6 mL 2.5 cm2
07201350 5 kDa 12/Pk. 20 mL 6 cm2
07201351 10 kDa 12/Pk. 20 mL 6 cm2
07201352 30 kDa 12/Pk. 20 mL 6 cm2
07201353 50 kDa 12/Pk. 20 mL 6 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 07201354 Supplier Corning™ Supplier No. 431491
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable, single use only ultrafiltration devices with polyethersulfone membranes (PES) for the centrifugal concentration and/or purification of biological fluids and aqueous solutions

Spin-X UF centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery.

  • The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides fast, high speed concen­trat­ing, even with particle laden solutions.
  • Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5,000, 10,000, 30,000, 50,000, and 100,000 to meet all of your concentrating needs. Choose an MWCO half to a third smaller than the protein to be concentrated.
  • The MWCO and graduations are printed on the side of the concentrator tube to avoid mix-ups.
  • Spin-X UF 6 and 20 concentrators can be used with either swinging bucket or fixed angle rotors. Spin-X UF 500 con­centrators require fixed angle rotors.

Specifications

Cap Material Polypropylene
Color White
Filter Type Centrifuge Filter
Material PES
Material (Membrane) PES
MWCO 100 kDa
Filtration Area 6 cm2
For Use With (Application) Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins, removal of labeled amino acids and nucleotides, HPLC sample preparation, deproteinization of samples, recovery of biomolecules from cell culture supernatants, lysates
Quantity 12/Pk.
Type Ultrafiltration Concentrator
Volume (English) Working 0.47 oz.
Volume (Metric) 20 mL
Volume (Metric) Working 14 mL
Height (English) 4.56 in.
Height (Metric) 11.6 cm
Diameter (English) 1.18 in.
Diameter (Metric) 3 cm
For Use With (Equipment) Swinging Bucket, Fixed Angle Rotor
Show More Show Less