missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FyterTech Nonwovens Spilfyter™KolorSafe™ Dry Base Neutralizers
Eliminate guesswork in neutralizing aggressive spills by going through three color changes to clearly indicate neutralization
$219.07 - $1228.09
Specifications
| Type | Dry Base Neutralizer |
|---|---|
| Format | Granular |
| For Use With (Application) | Common base spills |
| Includes | Color change indicator when neutralized |
| Packaging Quantity | 1 Ea. |
Description
- Provide a precise response to hazardous base spills such as ammonium hydroxide, potassium hydroxide, and sodium hydroxide
- Cost-effective formula is concentrated for easy application
- Ideal for outdoor leaks and spills
Specifications
| Dry Base Neutralizer | |
| Common base spills | |
| 1 Ea. |
| Granular | |
| Color change indicator when neutralized |