missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FyterTech Nonwovens Spilfyter™KolorSafe™ Dry Base Neutralizers
Description
- Provide a precise response to hazardous base spills such as ammonium hydroxide, potassium hydroxide, and sodium hydroxide
- Cost-effective formula is concentrated for easy application
- Ideal for outdoor leaks and spills
Specifications
Specifications
| Capacity (English) | 40 lb. |
| Capacity (Metric) | 18.14 kg |
| Type | Dry Base Neutralizer |
| Format | Granular |
| For Use With (Application) | Common base spills |
| Includes | Color change indicator when neutralized |
| Packaging Quantity | 1 Ea. |
| Weight (English) | 42 lb. |
| Weight (Metric) | 19 kg |