Learn More
FyterTech Nonwovens Spilfyter™ Hazmat Sorbents
Use for aggressive or unknown leaks or spills—offer a no-decision response
$84.87 - $465.71
Alert:
When picking up fuming acids, such as nitric and sulfuric, as well as strong oxidizers, care should be taken because they may degrade the sorbent material.
| Catalog Number | Mfr. No. | Capacity (English) | Color | Dimensions (L x W) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (English) | Color | Dimensions (L x W) | Price | Quantity | |||
|
19-066-618
|
FyterTech Nonwovens
S237 |
15 gal. | Yellow | 10 ft. x 3 in. |
Case of 6 Each for $158.34
|
|
|||
|
19-066-627
|
FyterTech Nonwovens
S296 |
66 gal. | Yellow | 300 ft. x 16 in. |
Case of 2 Each for $439.52
|
|
|||
|
19-812-543
|
FyterTech Nonwovens
NPSS230 |
15 gal. | Yellow | 4 ft. x 3 in. |
Case of 15 Each for $119.91
|
|
|||
|
29-635-1099
|
FyterTech Nonwovens
JVS271 |
10 gal. | Yellow | 12 x 12 in. |
Case of 100 Each for $84.87
|
|
|||
|
19-142-780
|
FyterTech Nonwovens
S261 |
10 gal. | White | 10 x 10 in. |
Case of 20 Each for $168.10
|
|
|||
|
19-066-617
|
FyterTech Nonwovens
S234 |
40 gal. | Yellow | 4 ft. x 3 in. |
Case of 40 Each for $360.92
|
|
|||
|
19-120-473
|
FyterTech Nonwovens
S266 |
10 gal. | White | 17 x 8.5 in. |
Case of 10 Each for $112.96
|
|
|||
|
19-066-622
|
FyterTech Nonwovens
S275 |
26 gal. | Yellow | 18 x 16 in. |
Case of 100 Each for $204.19
|
|
|||
|
19-066-626
|
FyterTech Nonwovens
S295 |
66 gal. | Yellow | 300 ft. x 32 in. |
Case for $465.71
|
|
|||
|
19-066-619
|
FyterTech Nonwovens
S262 |
25 gal. | White | 18 x 18 in. |
Case of 10 Each for $174.66
|
|
|||
Description
Premium yellow sorbents offer greater durability plus easily recognized “caution” color
- Bonded every 4 in. (10.2cm)
- All pads perforated vertically for a better fit