Learn More
Sphingosine Kinase Substrate
Catalog No. PV5372
Encompass
Change view
Click to view available options
Quantity:
400nmol
Catalog No. Quantity
PV5372 400nmol
1 options
Catalog No. PV5372 Supplier Thermo Fisher Scientific Supplier No. PV5372
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sphingosine Kinase Substrate
Sphingosine substrate specifically optimized for use with the Sphingosine kinases (SPHK1 & SPHK2). For use with the Adapta™ Universal Kinase Assay (PV5099).
Specifications
Chromagen Quantity 400nmol
For Use With (Application) Pharma and biopharma, target and lead identification and validation, drug discovery and development, kinase biology, biochemical kinase assays, Adapta™ universal kinase assays
Quantity 400nmol
Substrate Type Kinase Substrate
Product Type Sphingosine Kinase Substrate
Storage Requirements –20°C
For Research Use Only. Not for use in diagnostic procedures.