missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spermidine, free base, 99.5%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0215206805
Specifications
| Spermidine free base | |
| 124-20-9 | |
| 0.925g/mL | |
| C7H19N3 | |
| 5 g | |
| ATHGHQPFGPMSJY-UHFFFAOYSA-N | |
| (4-aminobutyl)(3-aminopropyl)amine | |
| 1102 | |
| 145.25 | |
| Liquid |
| Cell Biology and Analysis | |
| Colorless | |
| ∽ 99% | |
| MFCD00008229 | |
| spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl | |
| NCCCCNCCCN | |
| 145.25 | |
| CHEBI:16610 | |
| 0.995 |
Chemical Identifiers
| 124-20-9 | |
| 145.25 | |
| ATHGHQPFGPMSJY-UHFFFAOYSA-N | |
| 1102 | |
| (4-aminobutyl)(3-aminopropyl)amine |
| C7H19N3 | |
| MFCD00008229 | |
| spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl | |
| CHEBI:16610 | |
| NCCCCNCCCN |
Safety and Handling
EINECSNumber : 204-689-0
Recommended Storage : Store at 4°C, dessicate.