missing translation for 'onlineSavingsMsg'
Learn More

Spermidine, free base, 99.5%, MP Biomedicals™

Catalog No. p-4404883
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15206801 1 g
ICN15206805 5 g
ICN15206825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15206801 Supplier MP Biomedicals Inc Supplier No. 0215206801
Only null left

Chemical Identifiers

CAS 124-20-9
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008229
InChI Key ATHGHQPFGPMSJY-UHFFFAOYSA-N
Synonym spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl
PubChem CID 1102
ChEBI CHEBI:16610
IUPAC Name (4-aminobutyl)(3-aminopropyl)amine
SMILES NCCCCNCCCN

Specifications

Color Colorless
Density 0.925g/mL
Assay Percent Range ∽ 99%
Quantity 1 g
Formula Weight 145.25
Percent Purity 0.995
Physical Form Liquid
Chemical Name or Material Spermidine free base, Cell Biology and Analysis
EINECSNumber 204-689-0
Recommended Storage Store at 4°C, dessicate.