accessibility menu, dialog, popup

UserName

CAS RN 124-20-9

IUPAC Name: (4-aminobutyl)(3-aminopropyl)amine
Synonyms: spermidine spermidin
Molecular Weight (g/mol): 145.25
Molecular Formula: C7H19N3
InChi Key: ATHGHQPFGPMSJY-UHFFFAOYSA-N
SMILES: NCCCCNCCCN
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (4)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (3)
  • (2)