missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectinomycin Sulfate, ≥98%, MP Biomedicals™
$151.06 - $375.08
Chemical Identifiers
| CAS | 23312-56-3 |
|---|---|
| Molecular Formula | C14H26N2O11S |
| Molecular Weight (g/mol) | 430.43 |
| MDL Number | MFCD00270185 |
| InChI Key | XGBFWQUQYQIFLB-MTTMTQIXSA-N |
| Synonym | U 18409E, antibiotic |
| ChEBI | CHEBI:9216 |
| IUPAC Name | (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,â¸]tetradecan-7-one; sulfuric acid |
| SMILES | OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15899301
|
MP Biomedicals Inc
0215899301 |
1 g |
Each for $151.06
|
|
|||||
|
ICN15899305
|
MP Biomedicals Inc
0215899305 |
5 g |
Each for $375.08
|
|
|||||
Chemical Identifiers
| 23312-56-3 | |
| 430.43 | |
| XGBFWQUQYQIFLB-MTTMTQIXSA-N | |
| CHEBI:9216 | |
| OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
| C14H26N2O11S | |
| MFCD00270185 | |
| U 18409E, antibiotic | |
| (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,â¸]tetradecan-7-one; sulfuric acid |
Specifications
| Spectinomycin Sulfate | |
| 23312-56-3 | |
| 1 g | |
| C14H26N2O11S | |
| U 18409E, antibiotic | |
| OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O | |
| 430.43 | |
| CHEBI:9216 | |
| Spectinomycin Sulfate |
| Powder | |
| White | |
| ≥98% | |
| MFCD00270185 | |
| XGBFWQUQYQIFLB-MTTMTQIXSA-N | |
| (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,â¸]tetradecan-7-one; sulfuric acid | |
| 64740 | |
| 430.4 |