accessibility menu, dialog, popup

UserName

H3CHN(R)(S)HO(S)HNCH3(R)O(R)OH(S)O(R)CH3OO(R)(S)OHOHSOHOO

CAS RN 23312-56-3

IUPAC Name: (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
Synonyms: spectinomycin
Molecular Weight (g/mol): 430.43
Molecular Formula: C14H26N2O11S
InChi Key: XGBFWQUQYQIFLB-MTTMTQIXSA-N
SMILES: OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results