missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin Sulfate, ≥98%, MP Biomedicals™

Catalog No. ICN15899301
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15899301 1 g
ICN15899305 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15899301 Supplier MP Biomedicals Inc Supplier No. 0215899301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23312-56-3 , 7732-18-5
Molecular Formula C14H26N2O11S
Molecular Weight (g/mol) 430.43
MDL Number MFCD00270185
InChI Key XGBFWQUQYQIFLB-MTTMTQIXSA-N
Synonym U 18409E, antibiotic
ChEBI CHEBI:9216
IUPAC Name (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
SMILES OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Type Spectinomycin Sulfate
Physical Form Powder
Color White
Quantity 1 g
Assay Percent Range ≥98%
PubChem CID 64740
Formula Weight 430.4
Percent Purity ≥98%
Grade Not Available
Chemical Name or Material Spectinomycin Sulfate