missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sparkleen™ 1 Detergent

Sparkleen 1 is a universal detergent for manual or hand washing laboratory glassware and equipment. This mild highly buffered product is an excellent soak cleaner for most materials. Glassware washed with Sparkleen 1 is truly sparkling clean, entirely free from washing film, even with the hardest water.

$28.96 - $762.57

Specifications

Form Powder
pH 9.5 to 10.2 (0.5%)
Appearance White
For Use With (Application) Removes Proteins, Fats, Oils, media, wax pencil marks, dried Blood and other organic-inorganic deposits from Glassware
View More Specs

Products 2
missing translation for 'productCode' Volume (English) Price Quantity & Availability  
missing translation for 'productCode' Volume (English) Price Quantity & Availability  
043204
View Documents Promotion Details
3.25 lb.
Each for $28.96
Case of 12 Each for $347.52
Only null left
 
043205
View Documents Promotion Details
100 lb.
Each for $762.57
Only null left
 
Description

Description

Sparkleen 1 is a universal detergent for manual or hand washing laboratory glassware and equipment. This mild highly buffered product is an excellent soak cleaner for most materials. Glassware washed with Sparkleen 1 is truly sparkling clean, entirely free from washing film, even with the hardest water.

Especially valuable in exacting titrations and analyses. Excellent for presterilization cleaning and as soak solution to loosen frozen glass joints.

  • For manual washing
  • Fast-dissolving powder
  • Free rinsing
  • Contains phosphates
  • Softens water, removes stains

Specifications

Specifications

Powder
9.5 to 10.2 (0.5%)
White
Removes Proteins, Fats, Oils, media, wax pencil marks, dried Blood and other organic-inorganic deposits from Glassware
Videos
SDS
Documents

Documents