missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Sparkleen™ 1 Detergent

Catalog No. 043205
Change view
Click to view available options
Volume (English):
3.25 lb.
100 lb.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
043205 100 lb.
043204 3.25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 043205

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sparkleen 1 is a universal detergent for manual or hand washing laboratory glassware and equipment. This mild highly buffered product is an excellent soak cleaner for most materials. Glassware washed with Sparkleen 1 is truly sparkling clean, entirely free from washing film, even with the hardest water.

Especially valuable in exacting titrations and analyses. Excellent for presterilization cleaning and as soak solution to loosen frozen glass joints.

  • For manual washing
  • Fast-dissolving powder
  • Free rinsing
  • Contains phosphates
  • Softens water, removes stains

Specifications

Form Powder
pH 9.5 to 10.2 (0.5%)
Appearance White
Composition Sodium tripolyphosphate 10-50%, Sodium Carbonate 10 to 25%, Sodium Dodecylbenzenesulfo
Content And Storage Keep container tightly closed. Store in a dry cool area. Expected Shelf life 1 to 2 years. Disposal-Flush small quantities with plenty of water to the sanitary sewer. Dispose of in accordance with all local, state, and federal regulations.
For Use With (Application) Removes Proteins, Fats, Oils, media, wax pencil marks, dried Blood and other organic-inorganic deposits from Glassware
Packaging Drum
Volume (English) 100 lb.