missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Span 80 (=Sorbitan Monooleate), TCI America™
$45.57 - $83.54
Chemical Identifiers
| CAS | 1338-43-8 |
|---|---|
| Molecular Formula | C24H44O6 |
| Molecular Weight (g/mol) | 428.61 |
| MDL Number | MFCD00080948 |
| InChI Key | NWGKJDSIEKMTRX-NZAQQJATSA-N |
| Synonym | Arlacel 80, Sorbitan Monooleate |
| PubChem CID | 70693544 |
| IUPAC Name | 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O |
Chemical Identifiers
| 1338-43-8 | |
| 428.61 | |
| NWGKJDSIEKMTRX-NZAQQJATSA-N | |
| 70693544 | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O |
| C24H44O6 | |
| MFCD00080948 | |
| Arlacel 80, Sorbitan Monooleate | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate |
Specifications
| Brown-Yellow | |
| Clear Liquid at 20°C | |
| MFCD00080948 | |
| NWGKJDSIEKMTRX-NZAQQJATSA-N | |
| 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl (9Z)-octadec-9-enoate | |
| 70693544 |
| 1338-43-8 | |
| C24H44O6 | |
| Arlacel 80, Sorbitan Monooleate | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O | |
| 428.61 | |
| Span 80 (=Sorbitan Monooleate) |
Safety and Handling
EINECSNumber : (8)-0063
RTECSNumber : WG2932400
TSCA : Yes