missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ 5mm O.D. Thin Walled High-Throughput Economy NMR Tubes

Designed for routine use in all NMR spectrometers, Wilmad Economy NMR Tubes fit tightly in any spinner turbine and have minimal NMR background.

$33.50 - $37.44

Specifications

Material Borosilicate Glass
Product Type NMR Economy Sample Tube
Diameter (Metric) Outer 4.947±0.019 mm
Field Strength 100MHz
For Use With (Application) Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only.
View More Specs

Alert:

Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.


Products 2
Catalog Number Mfr. No. Length (Metric) Model Price Quantity  
Catalog Number Mfr. No. Length (Metric) Model Price Quantity  
16-800-184
View Documents
Bel-Art™
660000084
203 mm Wilmad WG-5MM-ECONOMY-8-5
Pack of 5 Each for $37.44
Only null left
 
16-800-183
View Documents
Bel-Art™
660000083
178 mm Wilmad WG-5MM-ECONOMY-7-5
Pack of 5 Each for $33.50
Only null left
 
Description

Description

  • High quality 5mm O.D. economy sample tube
  • Best O.D. tolerance in the industry and a 30% thicker wall than any competitors' product
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
  • Fit tightly in any spinner turbine
  • Minimal NMR background
  • Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A
  • Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes
  • Manufactured in an ISO 9001:2008 USA facility
Specifications

Specifications

Borosilicate Glass
4.947±0.019 mm
Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only.
0.43 mm
NMR Economy Sample Tube
100MHz
Tube and disposable cap
5/Pk.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'