missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ 5mm O.D. Thin Walled High-Throughput Economy NMR Tubes

Catalog No. 16800184
Change view
Click to view available options
Length (Metric):
178 mm
203 mm
Model:
Wilmad WG-5MM-ECONOMY-7-5
Wilmad WG-5MM-ECONOMY-8-5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Model
16800184 203 mm Wilmad WG-5MM-ECONOMY-8-5
16800183 178 mm Wilmad WG-5MM-ECONOMY-7-5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.
Catalog No. 16800184 Supplier Bel-Art™ Supplier No. 660000084
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.

Designed for routine use in all NMR spectrometers, Wilmad Economy NMR Tubes fit tightly in any spinner turbine and have minimal NMR background.

  • High quality 5mm O.D. economy sample tube
  • Best O.D. tolerance in the industry and a 30% thicker wall than any competitors' product
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
  • Fit tightly in any spinner turbine
  • Minimal NMR background
  • Made from borosilicate glass; meets ASTM E438 Type 1 Class B standard, recognized as N51A
  • Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes
  • Manufactured in an ISO 9001:2008 USA facility

Specifications

Product Type NMR Economy Sample Tube
Length (Metric) 203 mm
Material Borosilicate Glass
Diameter (Metric) Outer 4.947±0.019 mm
Field Strength 100MHz
For Use With (Application) Economy tubes are recommended only for experiments with small organic molecules (molecular weight approx.500) at ambient temperature only.
Includes Tube and disposable cap
Length (English) 8 in.
Thickness (Metric) Wall 0.43 mm
Quantity 5/Pk.
Model Wilmad WG-5MM-ECONOMY-8-5
Show More Show Less