missing translation for 'onlineSavingsMsg'
Learn More

Bel Art™ Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0

Catalog No. 08647850
Change view
Click to view available options
Color:
Amber
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
08647850 Amber
0864750 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08647850 Supplier Bel Art™ Supplier No. F420710008
Get Up to $500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Constructed of Durastar™ co-polyester. Standard desiccator cabinets block 99% of UV light and are resistant to staining, crazing, and chemical attack.

  • Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Stack up to 3 units high to save space
  • Can be used with desiccant cartridges, F420450250, F420450200, F420480100

Specifications

Autoclavable Not Autoclavable
Color Amber
Cabinet Style Vertical
Material Copolyester
Product Type Desiccator
Door Style Left-hinged and gasketed
For Use With (Application) Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
Includes Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model 1.0
No. of Shelves 1
Resistance Chemicals, stains and fine cracks
Shelf Type Fixed Perforated
Volume (English) 0.75 cu. ft.
Dimensions (L x W) 16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English) 16.3 in.
Length (Metric) 41.4 cm
Width (English) 13.4 in.
Width (English) Door 8.9 in.
Width (Metric) 34.1 cm
Width (Metric) Door 22.6 cm
Height (English) 8.4 in.
Height (English) Opening 4.5 in.
Height (Metric) 21.3 cm
Depth (English) 16.3 in.
Depth (Metric) 41.4 cm
Show More Show Less