missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SOURCE™ 15S Chromatography Media
Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides
$624.11 - $1,552.30
Specifications
| For Use With (Application) | For High Resolution Polishing PurifiCatIon of Proteins, Peptides and Oligonucleotides |
|---|---|
| Stability | Stable in common Ion Exchange Buffers |
| Storage Requirements | 4°C to 30°C |
| Product Type | Source 15 S Media |
| Flow Rate | 400 cm/Hoμr |
Description
- Average particle size: 15μm
- Ligand: sulphonate group
- Matrix: polystyrene/divinylbenzene
- pH stability working range: 2 to 13
- Available in 10mL and 50mL bottles
Specifications
| For High Resolution Polishing PurifiCatIon of Proteins, Peptides and Oligonucleotides | |
| 4°C to 30°C | |
| 400 cm/Hoμr | |
| 3.93 in. | |
| Strong CatIon Exchanger | |
| Polystyrene/Divinylbenzene | |
| 20% Ethanol, 0.2 M Sodium Acetate |
| Stable in common Ion Exchange Buffers | |
| Source 15 S Media | |
| 15 μm | |
| 10 cm | |
| 100 kPa | |
| 1 to 14 | |
| Sulphonate Group |