missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SOURCE™ 15S Chromatography Media

Catalog No. 45002034
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002034 50 mL
45002030 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 45002034 Supplier Cytiva Supplier No. 17094401

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides

  • Average particle size: 15 μm
  • Ligand: sulphonate group
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 13
  • Available in 10 mL and 50 mL bottles

Specifications

Quantity 50 mL
Form Spherical Beads
For Use With (Application) For High Resolution Polishing PurifiCatIon of Proteins, Peptides and Oligonucleotides
Stability Stable in common Ion Exchange Buffers
Storage Requirements 4°C to 30°C
Product Type Source 15 S Media
Flow Rate 400 cm/Hoμr
Particle Size 15 μm
Height (English) Bed 3.93 in.
Height (Metric) Bed 10 cm
Ion Exchange Type Strong CatIon Exchanger
Max. Pressure 100 kPa
Matrix Polystyrene/Divinylbenzene
pH Range 1 to 14
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphonate Group
Format Bulk Media
Show More Show Less