missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sortilin Recombinant Rabbit Monoclonal Antibody (23GB4170)
Rabbit Recombinant Monoclonal Antibody
$656.94 - $1779.35
Specifications
| Antigen | Sortilin |
|---|---|
| Clone | 23GB4170 |
| Content And Storage | -20°C |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
Description
Sortilin Recombinant Monoclonal Antibody for Western Blot
Sortilin functions as a sorting receptor in the Golgi compartment and as a clearance receptor on the cell surface. Sortilin is required for protein transport from the Golgi apparatus to the lysosomes by a pathway that is independent of the mannose-6-phosphate receptor (M6PR). It is also required for protein transport from the Golgi apparatus to the endosomes. Sortilin promotes neuronal apoptosis by mediating endocytosis of the proapoptotic precursor forms of BDNF (proBDNF) and NGFB (proNGFB). Also, Sortilin acts as a receptor for neurotensin.Specifications
| Sortilin | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Sort1 | |
| 100 kDa NT receptor; 2900053A11Rik; AI852375; Glycoprotein 110; glycoprotein 95; Gp110; Gp95; LDLCQ6; mNTR3; neurotensin receptor 3; NT3; NTR3; Nts3; Ntsr3; Sort1; Sortilin; sortilin 1 | |
| Sort1 | |
| Primary | |
| Affinity Chromatography |
| 23GB4170 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99523 | |
| 6272 | |
| A synthesized peptide derived from Sortilin (650-770AA). | |
| Antibody |