missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol Solution, FCC, 70%, Spectrum™ Chemical

Catalog No. p-7158304
Change view
Click to view available options
Quantity:
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606945 6 x 500 mL Amber Glass Bottle
18606946 4 L Amber Glass Bottle
18606947 4 x 4 L Amber Glass Bottle
18606948 20 L Poly Drum
18606949 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606945 Supplier Spectrum Chemical Mfg Cor Supplier No. S1525500MLCS6
Only null left

S1525, 50-70-4, C6H14O6

Spectrum™ Chemical Sorbitol Solution, 70 Percent, FCC is used as a sugar substitute and bonding agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-70-4 , 7732-18-5
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 0.7, 0.3
pH 5.0 to 7.5
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 6 x 500 mL
Percent Purity 64%