missing translation for 'onlineSavingsMsg'
Learn More

Sorbitol, Powder, NF, 91-100.5%, Spectrum™ Chemical

Catalog No. 18606960
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606960 500 g Poly Bottle
18606961 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18606960 Supplier Spectrum Chemical Mfg Cor Supplier No. SO219500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

SO219, 50-70-4, C6H14O6

Spectrum™ Chemical Sorbitol, Powder, NF is used in the pharmaceutical industry as a sweetener or humectant. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 50-70-4
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 1
pH 3.5 to 7.0
Boiling Point 295°C (563°F)
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 500 g
Percent Purity 91 to 100.5%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sorbitol Powder