missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbitan Trioleate, Spectrum™ Chemical
SPA85, 26266-58-0, C60H108O8
$1018.09 - $20215.03
Chemical Identifiers
| CAS | 26266-58-0 |
|---|---|
| Molecular Formula | C60H108O8 |
| Molecular Weight (g/mol) | 957.52 |
| InChI Key | PRXRUNOAOLTIEF-XDTJCZEISA-N |
| IUPAC Name | 2-[(2R,3S,4R)-4-hydroxy-3-[(9Z)-octadec-9-enoyloxy]oxolan-2-yl]-2-[(9Z)-octadec-9-enoyloxy]ethyl (9Z)-octadec-9-enoate |
| SMILES | CCCCCCCC\C=C/CCCCCCCC(=O)OCC(OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@@H](O)[C@@H]1OC(=O)CCCCCCC\C=C/CCCCCCCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606941
|
Spectrum Chemical Mfg Cor
SPA854LTGL |
4 L | Amber Glass Bottle |
Each for $1,018.09
|
|
||||
|
18606942
|
Spectrum Chemical Mfg Cor
SPA8520LT |
20 L | Poly Drum |
Each for $3,658.11
|
|
||||
|
18606943
|
Spectrum Chemical Mfg Cor
SPA85200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sorbitan TrioleateChemical Identifiers
| 26266-58-0 | |
| 957.52 | |
| 2-[(2R,3S,4R)-4-hydroxy-3-[(9Z)-octadec-9-enoyloxy]oxolan-2-yl]-2-[(9Z)-octadec-9-enoyloxy]ethyl (9Z)-octadec-9-enoate |
| C60H108O8 | |
| PRXRUNOAOLTIEF-XDTJCZEISA-N | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@@H](O)[C@@H]1OC(=O)CCCCCCC\C=C/CCCCCCCC |
Specifications
| 26266-58-0 | |
| Amber Glass Bottle | |
| 4 L | |
| CCCCCCCC\C=C/CCCCCCCC(=O)OCC(OC(=O)CCCCCCC\C=C/CCCCCCCC)[C@H]1OC[C@@H](O)[C@@H]1OC(=O)CCCCCCC\C=C/CCCCCCCC | |
| 957.52 | |
| Reagent |
| 1 | |
| C60H108O8 | |
| PRXRUNOAOLTIEF-XDTJCZEISA-N | |
| 2-[(2R,3S,4R)-4-hydroxy-3-[(9Z)-octadec-9-enoyloxy]oxolan-2-yl]-2-[(9Z)-octadec-9-enoyloxy]ethyl (9Z)-octadec-9-enoate | |
| Approx. 210 cps |