missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbic Acid, Powder, NF, 99-101%, Spectrum™ Chemical
SO215, 110-44-1, C6H8O2
$490.88 - $4745.79
Chemical Identifiers
| CAS | 110-44-1 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.13 |
| MDL Number | MFCD00002703 |
| InChI Key | WSWCOQWTEOXDQX-MQQKCMAXSA-N |
| IUPAC Name | (2E,4E)-hexa-2,4-dienoic acid |
| SMILES | C\C=C\C=C\C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606916
|
Spectrum Chemical Mfg Cor
SO215500GM |
500 g | Amber Glass Bottle |
Each for $490.88
|
|
||||
|
18606917
|
Spectrum Chemical Mfg Cor
SO21525KG |
2.5 kg | Amber Glass Bottle |
Each for $1,617.61
|
|
||||
|
18606918
|
Spectrum Chemical Mfg Cor
SO21512KG |
12 kg | Amber Glass Bottle |
Each for $4,745.79
|
|
||||
Description
Spectrum™ Chemical Sorbic Acid, Powder, NF is used as an antimicrobial agent in pharmaceutical formulations. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 110-44-1 | |
| 112.13 | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| C\C=C\C=C\C(O)=O |
| C6H8O2 | |
| MFCD00002703 | |
| (2E,4E)-hexa-2,4-dienoic acid |
Specifications
| 110-44-1 | |
| 0.002 | |
| Amber Glass Bottle | |
| MFCD00002703 | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| (2E,4E)-hexa-2,4-dienoic acid | |
| 99 to 101% | |
| Crystalline Powder or Lumps |
| 1 | |
| 132-135°C | |
| C6H8O2 | |
| 500 g | |
| C\C=C\C=C\C(O)=O | |
| 112.13 | |
| NF | |
| Sorbic Acid |