missing translation for 'onlineSavingsMsg'
Learn More

Sorbic Acid, Powder, NF, 99-101%, Spectrum™ Chemical

Catalog No. 18606916
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606916 500 g Amber Glass Bottle
18606917 2.5 kg Amber Glass Bottle
18606918 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606916 Supplier Spectrum Chemical Mfg Cor Supplier No. SO215500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

SO215, 110-44-1, C6H8O2

Spectrum™ Chemical Sorbic Acid, Powder, NF is used as an antimicrobial agent in pharmaceutical formulations. Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00002703
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.002
Melting Range 132-135°C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sorbic Acid