missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sorbic Acid, Powder, FCC, 99-101%, Spectrum™ Chemical
S1517, 110-44-1, C6H8O2
$510.06 - $14768.58
Chemical Identifiers
| CAS | 110-44-1 |
|---|---|
| Molecular Formula | C6H8O2 |
| Molecular Weight (g/mol) | 112.13 |
| MDL Number | MFCD00002703 |
| InChI Key | WSWCOQWTEOXDQX-MQQKCMAXSA-N |
| IUPAC Name | (2E,4E)-hexa-2,4-dienoic acid |
| SMILES | C\C=C\C=C\C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606911
|
Spectrum Chemical Mfg Cor
S1517500GM |
500 g | Amber Glass Bottle |
Each for $510.06
|
|
||||
|
18606912
|
Spectrum Chemical Mfg Cor
S151725KG |
2.5 kg | Amber Glass Bottle |
Each for $1,609.55
|
|
||||
|
18606913
|
Spectrum Chemical Mfg Cor
S151712KG |
12 kg | Amber Glass Bottle |
Each for $4,746.61
|
|
||||
|
18606914
|
Spectrum Chemical Mfg Cor
S151750KGBL |
50 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sorbic Acid, Powder, FCC is used as a food preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 110-44-1 | |
| 112.13 | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| C\C=C\C=C\C(O)=O |
| C6H8O2 | |
| MFCD00002703 | |
| (2E,4E)-hexa-2,4-dienoic acid |
Specifications
| 110-44-1 | |
| 0.002 | |
| Amber Glass Bottle | |
| MFCD00002703 | |
| WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
| (2E,4E)-hexa-2,4-dienoic acid | |
| 99 to 101% | |
| Crystalline Powder or Lumps |
| 1 | |
| 132-135°C | |
| C6H8O2 | |
| 500 g | |
| C\C=C\C=C\C(O)=O | |
| 112.13 | |
| FCC | |
| Sorbic Acid |