missing translation for 'onlineSavingsMsg'
Learn More

Sorbic Acid, Powder, FCC, 99-101%, Spectrum™ Chemical

Catalog No. 18606911
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606911 500 g Amber Glass Bottle
18606912 2.5 kg Amber Glass Bottle
18606913 12 kg Amber Glass Bottle
18606914 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18606911 Supplier Spectrum Chemical Mfg Cor Supplier No. S1517500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

S1517, 110-44-1, C6H8O2

Spectrum™ Chemical Sorbic Acid, Powder, FCC is used as a food preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00002703
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.002
Melting Range 132-135°C
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 99 to 101%
Grade FCC
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Sorbic Acid