Learn More
Sonoco™ ThermoSafe InsulatedShipper-PUR, Packed in Case
High performance polyurethane containers meet the needs of the most demanding temperature sensitive payloads
$525.03 - $626.94
| Catalog Number | Mfr. No. | Dimensions (L x W x H) Exterior | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W x H) Exterior | Price | Quantity & Availability | |||||
|
03532132
|
Sonoco™
E65UPS |
47.6 x 32.4 x 43.8 cm |
Pack of 2 Each for $525.03
|
|
|||||
|
03532150
|
Sonoco™
E50UPS |
37.4 x 35.2 x 59.05 cm |
Pack of 2 Each for $626.94
|
|
|||||
Description
Polyurethane insulation offers greater insulating power than equivalent thicknesses of EPS containers. Thick walls and 200 lb. outer corrugate offer protective shock absorption and cushioning during shipping and handling.
- Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
- Provides extensive protection for long range journeys
- Durable one-piece closed-cell polyurethane foam insulated shippers are CFC and HCFC-free—comply with international clean air regulations
- Proprietary process laminates highly efficient insulation core to inner and outer corrugate walls
- Withstands crush compression in excess of 5000 lb.
- Flexible polyurethane foam plug seals the top, cushions and insulates contents, vents dry ice
- R-value is +1.5 times EPS R-value, per inch of thickness
Specifications
| Polyurethane |